siRNA Details
Gene Symbol: TMEM135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168724.1 | NP_001162195.1 | 1 | 240 | ||
NM_022918.3 | NP_075069.3 | 1 | 240 | |||
NR_033149.1 | 1 | 240 | ||||
XM_017018140.1 | XP_016873629.1 | 1 | 233 | |||
GenBank mRNA | AK291804.1 | - | 1 | 150 | ||
AK302473.1 | - | 1 | 44 | |||
BC030952.1 | - | 1 | 99 | |||
BC051462.1 | - | 1 | 61 | 5278478 | ||
BX648678.1 | - | 1 | 226 | |||
DB469024.1 | - | 1 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
