siRNA Details
Gene Symbol: TMEM135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168724.1 | NP_001162195.1 | 2 | 418 | ||
NM_022918.3 | NP_075069.3 | 2 | 418 | |||
NR_033149.1 | 2 | 418 | ||||
GenBank mRNA | AK025757.1 | - | 1 | 32 | ||
AK225364.1 | - | 1 | 32 | |||
AK291804.1 | - | 2 | 328 | |||
AK302473.1 | - | 2 | 222 | |||
BC030952.1 | - | 2 | 277 | |||
BC051462.1 | - | 2 | 239 | 5278478 | ||
BX648678.1 | - | 2 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
