siRNA Details
Gene Symbol: RPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289111.1 | NP_001276040.1 | 1 | 86 | ||
NM_032194.2 | NP_115570.1 | 1 | 86 | |||
GenBank mRNA | AK024740.1 | - | 1 | 63 | ||
AK292165.1 | - | 1 | 65 | |||
AK301662.1 | - | 1 | 55 | |||
AL833613.1 | - | 1 | 54 | |||
BC035314.1 | - | 1 | 15 | 4745524 | ||
BG529605.1 | - | 1 | 87 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
