siRNA Details
Gene Symbol: C4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252204.1 | NP_001239133.1 | 1 | 38 | ||
NM_007293.2 | NP_009224.2 | 1 | 38 | |||
GenBank mRNA | AB209989.1 | - | 1 | 27 | ||
AK293191.1 | - | 1 | 38 | |||
AK293364.1 | - | 1 | 20 | |||
BC063289.1 | - | 1 | 71 | 5289068 | ||
BC146673.1 | - | 1 | 27 | 40148477 | ||
BC151204.1 | - | 1 | 25 | 40147083 | ||
DA304049.1 | - | 1 | 38 |
Gene Symbol: C4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002029.3 | NP_001002029.3 | 1 | 38 | ||
GenBank mRNA | AK304564.1 | - | 1 | 38 | ||
DA918359.1 | - | 1 | 38 |
Gene Symbol: C4B_2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242823.2 | NP_001229752.1 | 1 | 38 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
