siRNA Details
Gene Symbol: POTEJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511683.2 | XP_011509985.1 | 1 | 103 | ||
XM_017004741.1 | XP_016860230.1 | 2 | 76 |
Gene Symbol: POTEI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011511670.2 | XP_011509972.1 | 1 | 103 | ||
XM_017004732.1 | XP_016860221.1 | 2 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
