siRNA Details
Gene Symbol: SCRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031309.5 | NP_112599.2 | 2 | 2740 | ||
GenBank mRNA | AL512683.1 | - | 1 | 255 | ||
BC024967.2 | - | 1 | 261 | |||
BX420136.2 | - | 1 | 335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
