siRNA Details
Gene Symbol: PHOSPHO2-KLHL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199290.1 | NP_001186219.1 | 2 | 189 | ||
GenBank mRNA | AK090653.1 | - | 2 | 152 | ||
AX746559.1 | - | 2 | 152 |
Gene Symbol: PHOSPHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008489.3 | NP_001008489.1 | 2 | 189 | ||
NM_001199285.1 | NP_001186214.1 | 2 | 89 | |||
NM_001199286.1 | NP_001186215.1 | 2 | 308 | |||
NM_001199287.1 | NP_001186216.1 | 2 | 189 | |||
NM_001199288.1 | NP_001186217.1 | 2 | 189 | |||
GenBank mRNA | BC022324.2 | - | 1 | 20 | 4701753 | |
BC106013.1 | - | 2 | 178 | 4387304 | ||
BI550291.1 | - | 2 | 296 | |||
BM820383.1 | - | 2 | 156 | |||
DB443540.1 | - | 2 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
