siRNA Details
Gene Symbol: PHOSPHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008489.3 | NP_001008489.1 | 3 | 241 | ||
NM_001199285.1 | NP_001186214.1 | 3 | 141 | |||
NM_001199286.1 | NP_001186215.1 | 3 | 360 | |||
NM_001199287.1 | NP_001186216.1 | 3 | 241 | |||
GenBank mRNA | AK314915.1 | - | 3 | 144 | ||
BC022324.2 | - | 2 | 72 | 4701753 | ||
BC106013.1 | - | 3 | 230 | 4387304 | ||
BI550291.1 | - | 3 | 348 | |||
BM820383.1 | - | 3 | 208 | |||
DA211528.1 | - | 3 | 144 |
Gene Symbol: PHOSPHO2-KLHL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199290.1 | NP_001186219.1 | 3 | 241 | ||
GenBank mRNA | AK090653.1 | - | 3 | 204 | ||
AX746559.1 | - | 3 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
