siRNA Details
Gene Symbol: PHOSPHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008489.3 | NP_001008489.1 | 3 | 334 | ||
NM_001199285.1 | NP_001186214.1 | 3 | 234 | |||
NM_001199286.1 | NP_001186215.1 | 3 | 453 | |||
NM_001199287.1 | NP_001186216.1 | 3 | 334 | |||
GenBank mRNA | AK314915.1 | - | 3 | 237 | ||
BC022324.2 | - | 2 | 165 | 4701753 | ||
BC106013.1 | - | 3 | 323 | 4387304 | ||
BI550291.1 | - | 3 | 441 | |||
BM820383.1 | - | 3 | 301 | |||
DA211528.1 | - | 3 | 237 |
Gene Symbol: PHOSPHO2-KLHL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199290.1 | NP_001186219.1 | 3 | 334 | ||
GenBank mRNA | AK090653.1 | - | 3 | 297 | ||
AX746559.1 | - | 3 | 297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
