siRNA Details
Gene Symbol: RPL23AP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_000029.3 | 1 | 67 | |||
NR_024528.1 | 1 | 67 | ||||
NR_024529.1 | 1 | 67 | ||||
NR_024530.1 | 1 | 67 | ||||
NR_024531.1 | 1 | 67 | ||||
GenBank mRNA | BC000596.1 | - | 1 | 18 | ||
BC105600.1 | - | 1 | 19 | |||
BI464848.1 | - | 1 | 71 | |||
BX643196.1 | - | 1 | 41 | |||
DA864496.1 | - | 1 | 67 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
