siRNA Details
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026982.1 | 1 | 131 | |||
GenBank mRNA | AL365511.1 | - | 1 | 63 | ||
BC019348.1 | - | 1 | 48 |
Gene Symbol: RPL23AP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024528.1 | 1 | 96 | |||
NR_024529.1 | 1 | 96 | ||||
NR_024530.1 | 1 | 96 | ||||
GenBank mRNA | BG335018.1 | - | 1 | 40 | ||
BI464848.1 | - | 1 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
