siRNA Details
Gene Symbol: C14orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102366.2 | NP_001095836.1 | 5 | 759 | ||
NM_001102367.2 | NP_001095837.1 | 4 | 525 | |||
NM_001102368.2 | NP_001095838.1 | 3 | 343 | |||
NM_001102369.2 | NP_001095839.1 | 5 | 514 | |||
NM_001286470.1 | NP_001273399.1 | 6 | 1038 | |||
NM_001286471.1 | NP_001273400.1 | 3 | 594 | |||
NM_001286472.1 | NP_001273401.1 | 5 | 514 | |||
NM_001286473.1 | NP_001273402.1 | 5 | 514 | |||
NM_024952.7 | NP_079228.4 | 3 | 588 | |||
GenBank mRNA | AB210031.1 | - | 3 | 244 | ||
AK022564.1 | - | 3 | 159 | |||
AK023200.1 | - | 5 | 312 | |||
AK091394.1 | - | 3 | 162 | |||
AK097294.1 | - | 3 | 293 | |||
AK098752.1 | - | 3 | 294 | |||
AK123532.1 | - | 6 | 681 | |||
AK308857.1 | - | 2 | 205 | |||
AL602721.1 | - | 3 | 538 | |||
AY358516.1 | - | 3 | 594 | |||
BC009182.2 | - | 5 | 440 | |||
BC010614.2 | - | 4 | 474 | 4157432 | ||
BC065558.1 | - | 5 | 382 | 6138321 | ||
BX161393.1 | - | 3 | 272 | |||
BX247989.1 | - | 5 | 442 | |||
BX248060.1 | - | 3 | 397 | |||
BX248782.1 | - | 4 | 293 | |||
BX537970.1 | - | 3 | 290 | |||
DA296867.1 | - | 3 | 343 | |||
DA641301.1 | - | 5 | 462 | |||
DB258432.1 | - | 3 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
