siRNA Details
Gene Symbol: C14orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102366.2 | NP_001095836.1 | 3, 4 | 698 | ||
NM_001102367.2 | NP_001095837.1 | 2, 3 | 464 | |||
NM_001102368.2 | NP_001095838.1 | 1, 2 | 282 | |||
NM_001102369.2 | NP_001095839.1 | 3, 4 | 453 | |||
NM_001286470.1 | NP_001273399.1 | 4, 5 | 977 | |||
NM_001286471.1 | NP_001273400.1 | 1, 2 | 533 | |||
NM_001286472.1 | NP_001273401.1 | 3, 4 | 453 | |||
NM_001286473.1 | NP_001273402.1 | 3, 4 | 453 | |||
NM_024952.7 | NP_079228.4 | 1, 2 | 527 | |||
GenBank mRNA | AB210031.1 | - | 1, 2 | 183 | ||
AK022564.1 | - | 1, 2 | 98 | |||
AK023200.1 | - | 3, 4 | 251 | |||
AK091394.1 | - | 1, 2 | 101 | |||
AK097294.1 | - | 1, 2 | 232 | |||
AK098752.1 | - | 1, 2 | 233 | |||
AK123532.1 | - | 4, 5 | 620 | |||
AL602721.1 | - | 1, 2 | 477 | |||
AY358516.1 | - | 1, 2 | 533 | |||
BC009182.2 | - | 3, 4 | 379 | |||
BC010614.2 | - | 2, 3 | 413 | 4157432 | ||
BC065558.1 | - | 3, 4 | 321 | 6138321 | ||
BX161393.1 | - | 1, 2 | 211 | |||
BX247989.1 | - | 3, 4 | 381 | |||
BX248060.1 | - | 1, 2 | 336 | |||
BX248782.1 | - | 2, 3 | 232 | |||
BX537970.1 | - | 1, 2 | 229 | |||
DA296867.1 | - | 1, 2 | 282 | |||
DA641301.1 | - | 3, 4 | 401 | |||
DB258432.1 | - | 1, 2 | 371 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
