siRNA Details
Gene Symbol: C14orf159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102366.2 | NP_001095836.1 | 4 | 730 | ||
NM_001102367.2 | NP_001095837.1 | 3 | 496 | |||
NM_001102368.2 | NP_001095838.1 | 2 | 314 | |||
NM_001102369.2 | NP_001095839.1 | 4 | 485 | |||
NM_001286470.1 | NP_001273399.1 | 5 | 1009 | |||
NM_001286471.1 | NP_001273400.1 | 2 | 565 | |||
NM_001286472.1 | NP_001273401.1 | 4 | 485 | |||
NM_001286473.1 | NP_001273402.1 | 4 | 485 | |||
NM_024952.7 | NP_079228.4 | 2 | 559 | |||
GenBank mRNA | AB210031.1 | - | 2 | 215 | ||
AK022564.1 | - | 2 | 130 | |||
AK023200.1 | - | 4 | 283 | |||
AK091394.1 | - | 2 | 133 | |||
AK097294.1 | - | 2 | 264 | |||
AK098752.1 | - | 2 | 265 | |||
AK123532.1 | - | 5 | 652 | |||
AL602721.1 | - | 2 | 509 | |||
AY358516.1 | - | 2 | 565 | |||
BC009182.2 | - | 4 | 411 | |||
BC010614.2 | - | 3 | 445 | 4157432 | ||
BC065558.1 | - | 4 | 353 | 6138321 | ||
BX161393.1 | - | 2 | 243 | |||
BX247989.1 | - | 4 | 413 | |||
BX248060.1 | - | 2 | 368 | |||
BX248782.1 | - | 3 | 264 | |||
BX537970.1 | - | 2 | 261 | |||
DA296867.1 | - | 2 | 314 | |||
DA641301.1 | - | 4 | 433 | |||
DB258432.1 | - | 2 | 403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
