siRNA Details
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 2 | 239 | ||
NM_001322003.1 | NP_001308932.1 | 2 | 234 | |||
NM_001322005.1 | NP_001308934.1 | 2 | 351 | |||
NM_001322006.1 | NP_001308935.1 | 2 | 239 | |||
NM_001322007.1 | NP_001308936.1 | 2 | 239 | |||
NM_001322009.1 | NP_001308938.1 | 2 | 346 | |||
NM_001322011.1 | NP_001308940.1 | 2 | 239 | |||
NM_001322012.1 | NP_001308941.1 | 2 | 234 | |||
NM_001322013.1 | NP_001308942.1 | 2 | 234 | |||
NM_001322014.1 | NP_001308943.1 | 2 | 239 | |||
NM_001322015.1 | NP_001308944.1 | 2 | 234 | |||
NR_136154.1 | 2 | 239 | ||||
XM_017012342.1 | XP_016867831.1 | 2 | 213 | |||
GenBank mRNA | AB103082.1 | - | 2 | 152 | ||
AB103083.1 | - | 2 | 152 | |||
AB103085.1 | - | 1 | 152 | |||
AK312390.1 | - | 2 | 188 | |||
BC008400.1 | - | 2 | 197 | |||
BC031832.1 | - | 2 | 203 | |||
BC093921.1 | - | 2 | 174 | 7939766 | ||
BX537558.1 | - | 2 | 353 | |||
CR981250.1 | - | 2 | 140 | |||
U14658.1 | - | 2 | 154 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 7 | 1224 | |||
GenBank mRNA | U38980.1 | - | 7 | 1110 |
Gene Symbol: LOC105375355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250612.3 | XP_005250669.1 | 5 | 2889 | ||
XM_005250613.4 | XP_005250670.2 | 4 | 898 | |||
XM_006716140.3 | XP_006716203.1 | 7 | 2561 | |||
XM_006716141.3 | XP_006716204.1 | 6 | 2481 | |||
XM_017012885.1 | XP_016868374.1 | 4 | 898 | |||
XM_017012886.1 | XP_016868375.1 | 4 | 898 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 2 | 163 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 2 | 599 | |||
GenBank mRNA | AB017004.1 | - | 2 | 599 | ||
BC067809.1 | - | 2 | 231 | |||
BC110395.1 | - | 2 | 207 | |||
D38435.1 | - | 2 | 202 | |||
D38499.1 | - | 2 | 202 |
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 5 | 985 | |||
GenBank mRNA | BC101598.1 | - | 4 | 436 | 8069104 | |
D38437.1 | - | 1 | 134 | |||
D38498.1 | - | 1 | 134 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 2 | 208 | |||
NR_027776.2 | 2 | 208 | ||||
NR_027777.2 | 2 | 208 | ||||
GenBank mRNA | AK309660.1 | - | 2 | 156 | ||
BC052588.1 | - | 2 | 211 | |||
BF984173.1 | - | 2 | 207 | |||
D38436.1 | - | 2 | 189 | |||
D38439.1 | - | 1 | 120 | |||
D38501.1 | - | 1 | 120 | |||
D38502.1 | - | 2 | 189 | |||
DW407867.1 | - | 2 | 225 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 3 | 361 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 2 | 224 | |||
GenBank mRNA | AB017006.1 | - | 2 | 319 | ||
DB472687.1 | - | 2 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
