siRNA Details
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 2 | 103 | |||
NR_027776.2 | 2 | 103 | ||||
NR_027777.2 | 2 | 103 | ||||
GenBank mRNA | AK309660.1 | - | 2 | 51 | ||
BC052588.1 | - | 2 | 106 | |||
BF984173.1 | - | 2 | 102 | |||
D38436.1 | - | 2 | 84 | |||
D38439.1 | - | 1 | 15 | |||
D38501.1 | - | 1 | 15 | |||
D38502.1 | - | 2 | 84 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 7 | 1119 | |||
GenBank mRNA | DR007973.1 | - | 2 | 283 | ||
U38980.1 | - | 7 | 1005 |
Gene Symbol: LOC105375355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250612.3 | XP_005250669.1 | 5 | 2784 | ||
XM_005250613.4 | XP_005250670.2 | 4 | 793 | |||
XM_005250614.2 | XP_005250671.1 | 4 | 788 | |||
XM_006716140.3 | XP_006716203.1 | 7 | 2456 | |||
XM_006716141.3 | XP_006716204.1 | 6 | 2376 | |||
XM_017012885.1 | XP_016868374.1 | 4 | 793 | |||
XM_017012886.1 | XP_016868375.1 | 4 | 793 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 2 | 58 |
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 2 | 134 | ||
NM_001322003.1 | NP_001308932.1 | 2 | 129 | |||
NM_001322005.1 | NP_001308934.1 | 2 | 246 | |||
NM_001322006.1 | NP_001308935.1 | 2 | 134 | |||
NM_001322007.1 | NP_001308936.1 | 2 | 134 | |||
NM_001322009.1 | NP_001308938.1 | 2 | 241 | |||
NM_001322011.1 | NP_001308940.1 | 2 | 134 | |||
NM_001322012.1 | NP_001308941.1 | 2 | 129 | |||
NM_001322013.1 | NP_001308942.1 | 2 | 129 | |||
NM_001322014.1 | NP_001308943.1 | 2 | 134 | |||
NM_001322015.1 | NP_001308944.1 | 2 | 129 | |||
NR_136154.1 | 2 | 134 | ||||
XM_017012342.1 | XP_016867831.1 | 2 | 108 | |||
GenBank mRNA | AB103082.1 | - | 2 | 47 | ||
AB103083.1 | - | 2 | 47 | |||
AB103085.1 | - | 1 | 47 | |||
AB103087.1 | - | 2 | 47 | |||
AK312390.1 | - | 2 | 83 | |||
BC008400.1 | - | 2 | 92 | |||
BC031832.1 | - | 2 | 98 | |||
BC093921.1 | - | 2 | 69 | 7939766 | ||
BX537558.1 | - | 2 | 248 | |||
CR981250.1 | - | 2 | 35 | |||
DA774159.1 | - | 2 | 134 | |||
U14658.1 | - | 2 | 49 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 2 | 494 | |||
GenBank mRNA | AB017004.1 | - | 2 | 494 | ||
BC067809.1 | - | 2 | 126 | |||
BC110395.1 | - | 2 | 102 | |||
D38435.1 | - | 2 | 97 | |||
D38499.1 | - | 2 | 97 |
Gene Symbol: PMS2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047753.1 | - | 2 | 54 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 2 | 78 | |||
GenBank mRNA | DA215155.1 | - | 2 | 65 |
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 5 | 880 | |||
GenBank mRNA | BC046153.2 | - | 5 | 867 | ||
BC101598.1 | - | 4 | 331 | 8069104 | ||
BE938866.1 | - | 3 | 234 | |||
D38437.1 | - | 1 | 29 | |||
D38498.1 | - | 1 | 29 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 3 | 256 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 2 | 119 | |||
GenBank mRNA | AB017006.1 | - | 2 | 214 | ||
DB472687.1 | - | 2 | 119 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 1 | 495 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
