siRNA Details
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 8 | 1249 | |||
GenBank mRNA | BC101598.1 | - | 6 | 587 | 8069104 | |
D38437.1 | - | 4 | 397 | |||
D38498.1 | - | 4 | 397 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 9 | 1375 | |||
GenBank mRNA | U38980.1 | - | 9 | 1261 |
Gene Symbol: LOC105375355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012886.1 | XP_016868375.1 | 5 | 962 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 5 | 417 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 5 | 853 | |||
GenBank mRNA | AB017004.1 | - | 5 | 853 | ||
BC067809.1 | - | 5 | 485 | |||
BC110395.1 | - | 5 | 461 | |||
D38435.1 | - | 5 | 456 | |||
D38499.1 | - | 5 | 456 |
Gene Symbol: PMS2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022007.2 | 4 | 347 | |||
NR_046297.1 | 4 | 347 | ||||
NR_046299.1 | 4 | 347 | ||||
GenBank mRNA | BC029368.1 | - | 4 | 324 | ||
BC029419.1 | - | 4 | 312 | |||
CB141754.1 | - | 4 | 312 | |||
D38438.1 | - | 4 | 318 | |||
D38500.1 | - | 4 | 318 | |||
DA885243.1 | - | 4 | 348 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 5 | 462 | |||
NR_027776.2 | 4 | 359 | ||||
NR_027777.2 | 3 | 272 | ||||
GenBank mRNA | AK309660.1 | - | 4 | 307 | ||
BC052588.1 | - | 3 | 275 | |||
BF984173.1 | - | 4 | 358 | |||
D38436.1 | - | 5 | 443 | |||
D38439.1 | - | 3 | 271 | |||
D38501.1 | - | 3 | 271 | |||
D38502.1 | - | 5 | 443 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 11 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017006.1 | - | 5 | 583 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 3 | 750 | |||
GenBank mRNA | BM845848.1 | - | 4 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
