siRNA Details
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 3, 4 | 292 | ||
XM_005255738.4 | XP_005255795.1 | 3, 4 | 414 | |||
XM_006720830.3 | XP_006720893.1 | 3, 4 | 1312 | |||
XM_011522342.2 | XP_011520644.1 | 3, 4 | 341 | |||
XM_017022841.1 | XP_016878330.1 | 4, 5 | 365 | |||
XM_017022842.1 | XP_016878331.1 | 4, 5 | 417 | |||
XM_017022843.1 | XP_016878332.1 | 4, 5 | 478 | |||
XM_017022844.1 | XP_016878333.1 | 3, 4 | 285 | |||
XM_017022845.1 | XP_016878334.1 | 4, 5 | 365 | |||
XM_017022846.1 | XP_016878335.1 | 4, 5 | 365 | |||
XM_017022847.1 | XP_016878336.1 | 3, 4 | 305 | |||
GenBank mRNA | BQ101079.1 | - | 3, 4 | 338 |
Gene Symbol: LOC105376752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522797.2 | XP_011521099.1 | 3, 4 | 423 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 3, 4 | 292 | ||
XM_011522748.2 | XP_011521050.1 | 3, 4 | 389 | |||
XM_011522749.2 | XP_011521051.1 | 4, 5 | 501 | |||
XM_011522750.2 | XP_011521052.1 | 3, 4 | 415 | |||
XM_011522751.1 | XP_011521053.1 | 3, 4 | 388 | |||
XM_017023891.1 | XP_016879380.1 | 4, 5 | 501 | |||
GenBank mRNA | AF132984.1 | - | 3, 4 | 292 | ||
AK160377.1 | - | 22, 23 | 4479 | |||
BE253379.1 | - | 4, 5 | 560 | |||
KF175165.1 | - | 3, 4 | 328 | |||
KF175166.1 | - | 3, 4 | 328 | |||
KF175167.1 | - | 3, 4 | 328 | |||
KF175169.1 | - | 3, 4 | 292 | |||
KF175208.1 | - | 3, 4 | 439 | |||
KF175209.1 | - | 4, 5 | 510 | |||
KF175225.1 | - | 3, 4 | 292 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 3, 4 | 292 | ||
XM_005255489.3 | XP_005255546.1 | 5, 6 | 2711 | |||
XM_005255492.4 | XP_005255549.1 | 3, 4 | 696 | |||
XM_006720917.3 | XP_006720980.2 | 5, 6 | 2755 | |||
XM_011522587.1 | XP_011520889.1 | 5, 6 | 2755 | |||
XM_011522593.1 | XP_011520895.1 | 5, 6 | 2755 | |||
XM_011522594.2 | XP_011520896.1 | 5, 6 | 2755 | |||
XM_011522595.1 | XP_011520897.1 | 5, 6 | 2755 | |||
XM_011522596.1 | XP_011520898.1 | 3, 4 | 319 | |||
XM_011522597.1 | XP_011520899.1 | 5, 6 | 2755 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 3, 4 | 292 | ||
XM_005255484.3 | XP_005255541.1 | 4, 5 | 380 | |||
XM_005255485.4 | XP_005255542.1 | 3, 4 | 696 | |||
XM_011522576.2 | XP_011520878.2 | 5, 6 | 2745 | |||
XM_011522577.2 | XP_011520879.2 | 5, 6 | 2666 | |||
XM_011522578.2 | XP_011520880.2 | 3, 4 | 388 | |||
XM_011522580.2 | XP_011520882.2 | 3, 4 | 319 | |||
XM_011522581.1 | XP_011520883.1 | 5, 6 | 2755 | |||
XM_011522583.2 | XP_011520885.1 | 5, 6 | 2755 | |||
XM_011522584.1 | XP_011520886.1 | 5, 6 | 2755 | |||
XM_011522585.1 | XP_011520887.1 | 3, 4 | 319 | |||
XM_011522586.1 | XP_011520888.1 | 5, 6 | 2755 | |||
XM_017023553.1 | XP_016879042.1 | 5, 6 | 2755 | |||
XM_017023554.1 | XP_016879043.1 | 4, 5 | 2668 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 3, 4 | 292 | ||
XM_011522329.2 | XP_011520631.1 | 5, 6 | 703 | |||
XM_011522336.2 | XP_011520638.1 | 5, 6 | 696 | |||
XM_011522337.1 | XP_011520639.1 | 3, 4 | 319 | |||
GenBank mRNA | EL734608.1 | - | 3, 4 | 292 | ||
KF175168.1 | - | 3, 4 | 289 | |||
KF175170.1 | - | 3, 4 | 292 | |||
KF175171.1 | - | 3, 4 | 286 | |||
KF175172.1 | - | 3, 4 | 292 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 3, 4 | 292 | ||
XM_005255735.4 | XP_005255792.1 | 3, 4 | 414 | |||
XM_011522339.2 | XP_011520641.1 | 3, 4 | 340 | |||
XM_017022833.1 | XP_016878322.1 | 4, 5 | 365 | |||
XM_017022834.1 | XP_016878323.1 | 4, 5 | 417 | |||
XM_017022835.1 | XP_016878324.1 | 4, 5 | 478 | |||
XM_017022836.1 | XP_016878325.1 | 3, 4 | 285 | |||
XM_017022837.1 | XP_016878326.1 | 4, 5 | 365 | |||
XM_017022838.1 | XP_016878327.1 | 3, 4 | 760 | |||
XM_017022839.1 | XP_016878328.1 | 4, 5 | 365 | |||
XM_017022840.1 | XP_016878329.1 | 3, 4 | 305 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003610.1 | 22, 23 | 2374 | |||
GenBank mRNA | AL833251.1 | - | 22, 23 | 2374 | ||
BC063633.1 | - | 20, 21 | 1993 | |||
BC144350.1 | - | 3, 4 | 349 | 9052870 | ||
KF175207.1 | - | 3, 4 | 349 | |||
KF175221.1 | - | 3, 4 | 328 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 28, 29 | 5597 | |||
GenBank mRNA | AK124516.1 | - | 23, 24 | 3852 | ||
AK298381.1 | - | 1, 2 | 50 | |||
AK304604.1 | - | 8, 9 | 1297 | |||
BC039707.1 | - | 4, 5 | 611 | 4479933 | ||
BC046145.1 | - | 5, 6 | 703 | |||
KF175174.1 | - | 3, 4 | 328 | |||
KF175210.1 | - | 4, 5 | 571 | |||
KF175212.1 | - | 6, 7 | 461 | |||
KF175214.1 | - | 4, 5 | 510 | |||
KF175219.1 | - | 3, 4 | 478 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123721.1 | 22, 23 | 4385 | |||
NR_123722.1 | 12, 13 | 2246 | ||||
GenBank mRNA | AK131084.1 | - | 17, 18 | 4749 | ||
AK131085.1 | - | 16, 17 | 4655 | |||
AK302708.1 | - | 12, 13 | 2246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
