siRNA Details
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 4 | 575 | ||
XM_017023622.1 | XP_016879111.1 | 5 | 699 | |||
XM_017023623.1 | XP_016879112.1 | 5 | 699 | |||
XM_017023624.1 | XP_016879113.1 | 5 | 699 | |||
XM_017023625.1 | XP_016879114.1 | 5 | 679 | |||
XM_017023626.1 | XP_016879115.1 | 6 | 1318 | |||
XM_017023627.1 | XP_016879116.1 | 5 | 699 |
Gene Symbol: LOC105376752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522797.2 | XP_011521099.1 | 4 | 544 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 6 | 714 | ||
XM_017023514.1 | XP_016879003.1 | 6 | 1003 | |||
XM_017023515.1 | XP_016879004.1 | 7 | 1022 | |||
XM_017023516.1 | XP_016879005.1 | 6 | 1049 | |||
XM_017023517.1 | XP_016879006.1 | 6 | 780 | |||
XM_017023518.1 | XP_016879007.1 | 6 | 868 | |||
XM_017023519.1 | XP_016879008.1 | 6 | 925 | |||
XM_017023521.1 | XP_016879010.1 | 1 | 175 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 4 | 413 | ||
XM_011522748.2 | XP_011521050.1 | 4 | 510 | |||
XM_011522749.2 | XP_011521051.1 | 5 | 622 | |||
XM_011522750.2 | XP_011521052.1 | 4 | 536 | |||
XM_011522751.1 | XP_011521053.1 | 4 | 509 | |||
XM_017023891.1 | XP_016879380.1 | 5 | 622 | |||
GenBank mRNA | AF132984.1 | - | 4 | 413 | ||
AK160377.1 | - | 23 | 4600 | |||
KF175165.1 | - | 4 | 449 | |||
KF175166.1 | - | 4 | 449 | |||
KF175167.1 | - | 4 | 449 | |||
KF175209.1 | - | 5 | 631 | |||
KF175225.1 | - | 4 | 413 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277324.1 | NP_001264253.1 | 4 | 413 | ||
XM_005255489.3 | XP_005255546.1 | 6 | 2832 | |||
XM_005255492.4 | XP_005255549.1 | 4 | 817 | |||
XM_006720917.3 | XP_006720980.2 | 6 | 2876 | |||
XM_011522587.1 | XP_011520889.1 | 6 | 2876 | |||
XM_011522593.1 | XP_011520895.1 | 6 | 2876 | |||
XM_011522594.2 | XP_011520896.1 | 6 | 2876 | |||
XM_011522595.1 | XP_011520897.1 | 6 | 2876 | |||
XM_011522596.1 | XP_011520898.1 | 4 | 440 | |||
XM_011522597.1 | XP_011520899.1 | 6 | 2876 |
Gene Symbol: NPIPA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277323.1 | NP_001264252.1 | 4 | 413 | ||
XM_005255484.3 | XP_005255541.1 | 5 | 501 | |||
XM_005255485.4 | XP_005255542.1 | 4 | 817 | |||
XM_011522576.2 | XP_011520878.2 | 6 | 2866 | |||
XM_011522577.2 | XP_011520879.2 | 6 | 2787 | |||
XM_011522578.2 | XP_011520880.2 | 4 | 509 | |||
XM_011522580.2 | XP_011520882.2 | 4 | 440 | |||
XM_011522581.1 | XP_011520883.1 | 6 | 2876 | |||
XM_011522583.2 | XP_011520885.1 | 6 | 2876 | |||
XM_011522584.1 | XP_011520886.1 | 6 | 2876 | |||
XM_011522585.1 | XP_011520887.1 | 4 | 440 | |||
XM_011522586.1 | XP_011520888.1 | 6 | 2876 | |||
XM_017023553.1 | XP_016879042.1 | 6 | 2876 | |||
XM_017023554.1 | XP_016879043.1 | 5 | 2789 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 4 | 413 | ||
XM_011522329.2 | XP_011520631.1 | 6 | 824 | |||
XM_011522336.2 | XP_011520638.1 | 6 | 817 | |||
XM_011522337.1 | XP_011520639.1 | 4 | 440 | |||
GenBank mRNA | CB123100.1 | - | 1 | 91 | ||
EL734608.1 | - | 4 | 413 | |||
KF175168.1 | - | 4 | 410 | |||
KF175170.1 | - | 4 | 413 | |||
KF175171.1 | - | 4 | 407 | |||
KF175172.1 | - | 4 | 413 | |||
KF175173.1 | - | 2 | 188 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 4 | 413 | ||
XM_005255735.4 | XP_005255792.1 | 4 | 535 | |||
XM_011522339.2 | XP_011520641.1 | 4 | 461 | |||
XM_017022833.1 | XP_016878322.1 | 5 | 486 | |||
XM_017022834.1 | XP_016878323.1 | 5 | 538 | |||
XM_017022835.1 | XP_016878324.1 | 5 | 599 | |||
XM_017022836.1 | XP_016878325.1 | 4 | 406 | |||
XM_017022837.1 | XP_016878326.1 | 5 | 486 | |||
XM_017022838.1 | XP_016878327.1 | 4 | 881 | |||
XM_017022839.1 | XP_016878328.1 | 5 | 486 | |||
XM_017022840.1 | XP_016878329.1 | 4 | 426 | |||
GenBank mRNA | BI046418.1 | - | 1 | 64 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 4 | 413 | ||
XM_005255738.4 | XP_005255795.1 | 4 | 535 | |||
XM_006720830.3 | XP_006720893.1 | 4 | 1433 | |||
XM_011522342.2 | XP_011520644.1 | 4 | 462 | |||
XM_017022841.1 | XP_016878330.1 | 5 | 486 | |||
XM_017022842.1 | XP_016878331.1 | 5 | 538 | |||
XM_017022843.1 | XP_016878332.1 | 5 | 599 | |||
XM_017022844.1 | XP_016878333.1 | 4 | 406 | |||
XM_017022845.1 | XP_016878334.1 | 5 | 486 | |||
XM_017022846.1 | XP_016878335.1 | 5 | 486 | |||
XM_017022847.1 | XP_016878336.1 | 4 | 426 | |||
GenBank mRNA | BQ101079.1 | - | 4 | 459 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 4 | 521 | ||
GenBank mRNA | KF175195.1 | - | 4 | 425 | ||
KF175204.1 | - | 4 | 464 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 4 | 543 | ||
XM_011523496.2 | XP_011521798.1 | 5 | 599 | |||
XM_011523498.2 | XP_011521800.1 | 5 | 633 | |||
XM_017023248.1 | XP_016878737.1 | 5 | 1036 | |||
XM_017023249.1 | XP_016878738.1 | 4 | 853 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 5 | 597 | ||
GenBank mRNA | AB209632.1 | - | 4 | 3164 | ||
AK294452.1 | - | 1 | 160 | |||
AK295170.1 | - | 5 | 596 | |||
AK303500.1 | - | 1 | 91 | |||
AK304162.1 | - | 3 | 379 | |||
AK316459.1 | - | 1 | 26 | |||
BC036263.1 | - | 5 | 597 | 3903371 | ||
BC094882.1 | - | 4 | 524 | 3093162 | ||
BM557638.1 | - | 1 | 171 | |||
KF175175.1 | - | 4 | 464 | |||
KF175176.1 | - | 4 | 521 | |||
KF175177.1 | - | 4 | 521 | |||
KF175178.1 | - | 4 | 422 | |||
KF175179.1 | - | 4 | 464 | |||
KF175180.1 | - | 4 | 464 | |||
KF175181.1 | - | 4 | 608 | |||
KF175182.1 | - | 4 | 464 | |||
KF175183.1 | - | 3 | 356 | |||
KF175184.1 | - | 3 | 347 | |||
KF175185.1 | - | 4 | 464 | |||
KF175186.1 | - | 3 | 338 | |||
KF175187.1 | - | 4 | 464 | |||
KF175188.1 | - | 4 | 425 | |||
KF175189.1 | - | 4 | 425 | |||
KF175190.1 | - | 4 | 464 | |||
KF175191.1 | - | 4 | 425 | |||
KF175192.1 | - | 4 | 425 | |||
KF175193.1 | - | 4 | 428 | |||
KF175194.1 | - | 4 | 428 | |||
KF175196.1 | - | 4 | 425 | |||
KF175197.1 | - | 4 | 425 | |||
KF175198.1 | - | 4 | 521 | |||
KF175199.1 | - | 4 | 521 | |||
KF175200.1 | - | 4 | 521 | |||
KF175201.1 | - | 4 | 464 | |||
KF175202.1 | - | 4 | 425 | |||
KF175203.1 | - | 4 | 521 | |||
KF175205.1 | - | 4 | 521 | |||
KF175206.1 | - | 4 | 521 | |||
KF175215.1 | - | 6 | 783 | |||
KF175216.1 | - | 5 | 798 | |||
KF175218.1 | - | 4 | 426 | |||
KF175223.1 | - | 4 | 521 | |||
KF175224.1 | - | 4 | 464 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 5 | 746 | ||
XM_017023244.1 | XP_016878733.1 | 4 | 2757 | |||
XM_017023245.1 | XP_016878734.1 | 5 | 2594 | |||
XM_017023246.1 | XP_016878735.1 | 5 | 2515 | |||
XM_017023247.1 | XP_016878736.1 | 4 | 748 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 4 | 800 | ||
GenBank mRNA | AK296301.1 | - | 81 | |||
AK296338.1 | - | 4 | 641 | |||
AK303264.1 | - | 1 | 113 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 4 | 613 | ||
XM_005255741.3 | XP_005255798.1 | 5 | 1425 | |||
XM_005255742.4 | XP_005255799.1 | 5 | 1171 | |||
XM_011545929.2 | XP_011544231.1 | 6 | 1245 | |||
XM_011545931.2 | XP_011544233.1 | 5 | 878 | |||
XM_017023628.1 | XP_016879117.1 | 5 | 628 | |||
XM_017023629.1 | XP_016879118.1 | 5 | 1171 | |||
XM_017023630.1 | XP_016879119.1 | 6 | 898 | |||
XM_017023631.1 | XP_016879120.1 | 4 | 1242 | |||
XM_017023632.1 | XP_016879121.1 | 5 | 628 | |||
GenBank mRNA | BC144439.1 | - | 4 | 574 | ||
BC157836.1 | - | 4 | 1263 | 9057055 | ||
KF175222.1 | - | 4 | 575 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 4 | 613 | ||
NM_001287251.1 | NP_001274180.1 | 5 | 699 | |||
XM_003118719.7 | XP_003118767.2 | 5 | 1174 | |||
XM_011545690.2 | XP_011543992.1 | 6 | 1263 | |||
XM_011545693.2 | XP_011543995.1 | 6 | 1262 | |||
XM_017022814.1 | XP_016878303.1 | 6 | 1263 | |||
XM_017022815.1 | XP_016878304.1 | 6 | 783 | |||
XM_017022816.1 | XP_016878305.1 | 6 | 1263 | |||
XM_017022817.1 | XP_016878306.1 | 5 | 1175 | |||
XM_017022818.1 | XP_016878307.1 | 5 | 2500 | |||
XM_017022819.1 | XP_016878308.1 | 6 | 1190 | |||
XM_017022820.1 | XP_016878309.1 | 6 | 907 | |||
XM_017022821.1 | XP_016878310.1 | 5 | 1773 | |||
XM_017022822.1 | XP_016878311.1 | 6 | 2776 | |||
XM_017022823.1 | XP_016878312.1 | 4 | 2412 | |||
XM_017022824.1 | XP_016878313.1 | 6 | 1189 | |||
XM_017022825.1 | XP_016878314.1 | 6 | 907 | |||
XM_017022826.1 | XP_016878315.1 | 5 | 724 | |||
XM_017022828.1 | XP_016878317.1 | 6 | 1262 | |||
GenBank mRNA | BC171825.1 | - | 5 | 1349 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003610.1 | 23 | 2495 | |||
GenBank mRNA | AL833251.1 | - | 23 | 2495 | ||
BC063633.1 | - | 21 | 2114 | |||
BC144350.1 | - | 4 | 470 | 9052870 | ||
KF175207.1 | - | 4 | 470 | |||
KF175220.1 | - | 4 | 375 | |||
KF175221.1 | - | 4 | 449 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036447.1 | 29 | 5718 | |||
GenBank mRNA | AK124516.1 | - | 24 | 3973 | ||
AK298381.1 | - | 2 | 171 | |||
BC039707.1 | - | 5 | 732 | 4479933 | ||
BC046145.1 | - | 6 | 824 | |||
KF175174.1 | - | 4 | 449 | |||
KF175211.1 | - | 6 | 769 | |||
KF175212.1 | - | 7 | 582 | |||
KF175214.1 | - | 5 | 631 | |||
KF175219.1 | - | 4 | 599 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123722.1 | 13 | 2367 | |||
GenBank mRNA | AK131085.1 | - | 17 | 4776 | ||
AK302708.1 | - | 13 | 2367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
