siRNA Details
Gene Symbol: HBQ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005331.4 | NP_005322.1 | 1 | 208 | ||
GenBank mRNA | BC056686.1 | - | 1 | 77 | 6738551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
