siRNA Details
Gene Symbol: DDT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001084392.1 | NP_001077861.1 | 2 | 201 | ||
NM_001355.3 | NP_001346.1 | 3 | 252 | |||
GenBank mRNA | AK292044.1 | - | 2 | 145 | ||
BC005971.1 | - | 2 | 149 | 4082777 | ||
BC015508.2 | - | 2 | 170 | 3902943 | ||
BT006970.1 | - | 2 | 124 | |||
CR456431.1 | - | 2 | 147 | |||
U49785.1 | - | 3 | 250 | |||
U84143.1 | - | 2 | 138 |
Gene Symbol: DDTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001084393.1 | NP_001077862.1 | 2 | 201 | ||
XM_011529816.2 | XP_011528118.1 | 2 | 818 | |||
GenBank mRNA | AK296110.1 | - | 2 | 151 | ||
AK296189.1 | - | 2 | 151 | |||
AK298326.1 | - | 2 | 143 | |||
DB157727.1 | - | 2 | 201 |
Gene Symbol: GSTT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CN271767.1 | - | 1 | 505 |
Gene Symbol: LOC391322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144931.1 | NP_001138403.1 | 2 | 145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
