siRNA Details
Gene Symbol: RRN3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003370.2 | 2 | 94 | |||
GenBank mRNA | BC009198.2 | - | 3 | 324 | ||
BC068999.1 | - | 3 | 324 | |||
BX101439.1 | - | 2 | 184 | |||
CN397262.1 | - | 2 | 101 |
Gene Symbol: RRN3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003369.2 | 2 | 308 | |||
GenBank mRNA | AK225670.1 | - | 2 | 156 | ||
AK298442.1 | - | 2 | 131 | |||
AK301253.1 | - | 2 | 128 | |||
DC420905.1 | - | 2 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
