siRNA Details
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301064.1 | NP_001287993.1 | 3 | 320 | ||
NM_018427.4 | NP_060897.3 | 3 | 320 | |||
XM_005255376.1 | XP_005255433.1 | 3 | 229 | |||
XM_005255377.4 | XP_005255434.1 | 3 | 268 | |||
XM_011522541.1 | XP_011520843.1 | 3 | 229 | |||
GenBank mRNA | AF227156.1 | - | 3 | 286 | ||
AJ272050.1 | - | 3 | 240 | |||
AK225662.1 | - | 3 | 296 | |||
AK300253.1 | - | 3 | 229 | |||
AK304856.1 | - | 3 | 186 | |||
AK314769.1 | - | 3 | 234 | |||
AK316520.1 | - | 3 | 234 | |||
BC036182.1 | - | 3 | 235 | |||
BC104660.1 | - | 3 | 242 | |||
BC132688.1 | - | 3 | 237 | 40146710 | ||
BC132690.1 | - | 3 | 237 | 40146712 | ||
DC376629.1 | - | 3 | 321 |
Gene Symbol: RRN3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003370.2 | 3 | 195 | |||
GenBank mRNA | BC009198.2 | - | 4 | 425 | ||
BC068999.1 | - | 4 | 425 | |||
BX101439.1 | - | 3 | 285 | |||
CN397262.1 | - | 3 | 202 |
Gene Symbol: RRN3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003369.2 | 3 | 409 | |||
GenBank mRNA | AK225670.1 | - | 3 | 257 | ||
AK298442.1 | - | 3 | 232 | |||
AK301253.1 | - | 3 | 229 | |||
DC420905.1 | - | 3 | 432 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
