siRNA Details
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018427.4 | NP_060897.3 | 4 | 409 | ||
XM_005255376.1 | XP_005255433.1 | 4 | 318 | |||
XM_011522541.1 | XP_011520843.1 | 4 | 318 | |||
GenBank mRNA | AF227156.1 | - | 4 | 375 | ||
AJ272050.1 | - | 4 | 329 | |||
AK225662.1 | - | 4 | 385 | |||
AK302994.1 | - | 1 | 56 | |||
AK304856.1 | - | 4 | 275 | |||
AK314769.1 | - | 4 | 323 | |||
BC036182.1 | - | 4 | 324 | |||
BC104660.1 | - | 4 | 331 | |||
BC132688.1 | - | 4 | 326 | 40146710 | ||
BC132690.1 | - | 4 | 326 | 40146712 | ||
DC376629.1 | - | 4 | 410 |
Gene Symbol: RRN3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003370.2 | 4 | 284 | |||
GenBank mRNA | BC009198.2 | - | 5 | 514 | ||
BC068999.1 | - | 5 | 514 | |||
BX101439.1 | - | 4 | 374 | |||
CN397262.1 | - | 4 | 291 |
Gene Symbol: RRN3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003369.2 | 4 | 498 | |||
GenBank mRNA | AK225670.1 | - | 4 | 346 | ||
AK301253.1 | - | 4 | 318 | |||
DC420905.1 | - | 4 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
