siRNA Details
Gene Symbol: CYP4F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023944.3 | NP_076433.3 | 6 | 706 | ||
NR_117085.1 | 5 | 724 | ||||
XM_006722850.3 | XP_006722913.1 | 6 | 591 | |||
XM_011528202.2 | XP_011526504.1 | 4 | 226 | |||
XM_011528203.2 | XP_011526505.1 | 3 | 1448 | |||
XM_011528204.2 | XP_011526506.1 | 4 | 927 | |||
XM_011528205.2 | XP_011526507.1 | 3 | 139 | |||
XM_011528207.2 | XP_011526509.1 | 3 | 1835 | |||
XM_011528208.2 | XP_011526510.1 | 5 | 345 | |||
XM_017027172.1 | XP_016882661.1 | 6 | 648 | |||
GenBank mRNA | AB035130.1 | - | 6 | 617 | ||
AB035131.1 | - | 6 | 617 | |||
AK074412.1 | - | 1 | 950 | |||
AK075435.1 | - | 5 | 652 | |||
AL832171.1 | - | 6 | 559 | |||
AY008841.1 | - | 5 | 587 | |||
AY358977.1 | - | 6 | 628 | |||
BC035350.1 | - | 6 | 633 | 5187264 |
Gene Symbol: CYP4F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082.4 | NP_001073.3 | 6 | 683 | ||
GenBank mRNA | AK290790.1 | - | 6 | 637 | ||
AK296200.1 | - | 4 | 530 | |||
AK300961.1 | - | 5 | 489 | |||
AK313705.1 | - | 6 | 628 | |||
BC067437.1 | - | 6 | 624 | 7002207 | ||
BC067438.1 | - | 6 | 624 | |||
BC067439.1 | - | 6 | 624 | 7002212 | ||
BC067440.1 | - | 6 | 624 | 7002214 | ||
D26480.1 | - | 6 | 628 | |||
U02388.2 | - | 6 | 646 |
Gene Symbol: CYP4F24P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033864.1 | 5 | 773 | |||
GenBank mRNA | AK297837.1 | - | 5 | 668 | ||
AK308750.1 | - | 5 | 768 |
Gene Symbol: CYP4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000896.2 | NP_000887.2 | 6 | 637 | ||
NM_001199208.1 | NP_001186137.1 | 6 | 637 | |||
NM_001199209.1 | NP_001186138.1 | 6 | 621 | |||
XM_005259911.4 | XP_005259968.1 | 4 | 565 | |||
XM_011528014.2 | XP_011526316.1 | 4 | 414 | |||
XM_017026814.1 | XP_016882303.1 | 5 | 715 | |||
XM_017026815.1 | XP_016882304.1 | 5 | 720 | |||
GenBank mRNA | AB002454.1 | - | 6 | 637 | ||
AF054821.1 | - | 5 | 618 | |||
AK296761.1 | - | 6 | 556 | |||
AK296802.1 | - | 5 | 727 | |||
AK298675.1 | - | 6 | 617 | |||
AK304200.1 | - | 6 | 634 | |||
AK316136.1 | - | 6 | 621 | |||
BC136299.1 | - | 6 | 629 | 9020286 | ||
D12620.1 | - | 6 | 621 | |||
D12621.1 | - | 6 | 603 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001100187, NM_024442, AK078557, AK146232, AF233646, and others.
Gene Ontology Categories:
