siRNA Details
Gene Symbol: CYP4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000896.2 | NP_000887.2 | 12 | 1396 | ||
NM_001199208.1 | NP_001186137.1 | 12 | 1396 | |||
NM_001199209.1 | NP_001186138.1 | 12 | 1380 | |||
XM_005259911.4 | XP_005259968.1 | 10 | 1324 | |||
XM_011528014.2 | XP_011526316.1 | 10 | 1173 | |||
XM_017026814.1 | XP_016882303.1 | 11 | 1474 | |||
GenBank mRNA | AB002454.1 | - | 12 | 1396 | ||
AF054821.1 | - | 11 | 1377 | |||
AK296761.1 | - | 12 | 1315 | |||
AK296802.1 | - | 11 | 1486 | |||
AK298675.1 | - | 12 | 1376 | |||
AK303477.1 | - | 6 | 739 | |||
AK304200.1 | - | 12 | 1393 | |||
AK316136.1 | - | 12 | 1380 | |||
BC136299.1 | - | 12 | 1388 | 9020286 | ||
D12620.1 | - | 12 | 1380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
