siRNA Details
Gene Symbol: CEACAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277163.2 | NP_001264092.1 | 1, 2 | 158 | ||
NM_001815.4 | NP_001806.2 | 1, 2 | 158 | |||
NR_102333.2 | 1, 2 | 158 | ||||
GenBank mRNA | AK298694.1 | - | 1, 2 | 158 | ||
BC106728.1 | - | 1, 2 | 96 | 40015072 | ||
D90277.1 | - | 1, 2 | 144 | |||
D90278.1 | - | 1, 2 | 131 | |||
DA674909.1 | - | 1, 2 | 146 | |||
DC428993.1 | - | 1, 2 | 158 | |||
E03348.1 | - | 1, 2 | 144 | |||
E03349.1 | - | 1, 2 | 131 | |||
L00692.1 | - | 1, 2 | 124 | |||
L00693.1 | - | 1, 2 | 124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
