siRNA Details
Gene Symbol: CEACAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291484.2 | NP_001278413.1 | 2 | 401 | ||
NM_001308398.1 | NP_001295327.1 | 2 | 401 | |||
NM_004363.5 | NP_004354.3 | 2 | 401 | |||
XM_005258413.3 | XP_005258470.2 | 2 | 370 | |||
XM_011526322.1 | XP_011524624.1 | 2 | 370 | |||
XM_017026145.1 | XP_016881634.1 | 2 | 370 | |||
XM_017026146.1 | XP_016881635.1 | 2 | 370 | |||
GenBank mRNA | AB852566.1 | - | 2 | 254 | ||
AB852567.1 | - | 2 | 254 | |||
AK223101.1 | - | 2 | 337 | |||
AK310844.1 | - | 2 | 360 | |||
BC034671.1 | - | 2 | 349 | 5184800 | ||
BP272354.1 | - | 2 | 401 | |||
CR749337.1 | - | 2 | 289 | |||
DB006625.1 | - | 2 | 375 | |||
M15042.1 | - | 2 | 243 | |||
M29540.1 | - | 2 | 368 | |||
X16455.1 | - | 2 | 243 |
Gene Symbol: CEACAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277163.2 | NP_001264092.1 | 2 | 342 | ||
NM_001815.4 | NP_001806.2 | 2 | 342 | |||
NR_102333.2 | 2 | 342 | ||||
GenBank mRNA | AK298694.1 | - | 2 | 342 | ||
BC106728.1 | - | 2 | 280 | 40015072 | ||
D90277.1 | - | 2 | 328 | |||
DA674909.1 | - | 2 | 330 | |||
DC428993.1 | - | 2 | 342 | |||
E03348.1 | - | 2 | 328 | |||
L00692.1 | - | 2 | 308 | |||
L00693.1 | - | 2 | 308 |
Gene Symbol: CEACAM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002483.6 | NP_002474.4 | 2 | 373 | ||
XM_011526990.1 | XP_011525292.1 | 2 | 472 | |||
GenBank mRNA | AK312542.1 | - | 2 | 358 | ||
BC005008.1 | - | 2 | 294 | 3640231 | ||
BT009774.1 | - | 2 | 254 | |||
DA442875.1 | - | 2 | 474 | |||
M18216.1 | - | 2 | 300 | |||
M18728.1 | - | 2 | 304 | |||
M29541.1 | - | 2 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
