siRNA Details
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035511.1 | NP_001030588.1 | 5 | 331 | ||
NM_001035512.1 | NP_001030589.1 | 5 | 393 | |||
NM_001035513.1 | NP_001030590.1 | 4 | 336 | |||
NM_001278172.1 | NP_001265101.1 | 4 | 229 | |||
NM_003001.3 | NP_002992.1 | 6 | 495 | |||
NR_103459.1 | 7 | 522 | ||||
GenBank mRNA | AB201252.1 | - | 4 | 324 | ||
AB211234.1 | - | 5 | 387 | |||
AB211235.1 | - | 5 | 325 | |||
AB212048.1 | - | 4 | 217 | |||
AK131051.1 | - | 6 | 490 | |||
AK293687.1 | - | 5 | 385 | |||
AK294305.1 | - | 5 | 326 | |||
BC020808.1 | - | 6 | 490 | 4291847 | ||
BC033626.1 | - | 6 | 488 | 5416956 | ||
BC066329.1 | - | 6 | 490 | 5301907 | ||
CR457102.1 | - | 5 | 465 | |||
D49737.1 | - | 6 | 478 | |||
DA751501.1 | - | 7 | 507 | |||
KU178321.1 | - | 6 | 465 | |||
KU178322.1 | - | 2 | 306 | |||
KU178323.1 | - | 5 | 301 | |||
KU178324.1 | - | 4 | 244 | |||
U57877.1 | - | 6 | 492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
