siRNA Details
Gene Symbol: APOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000040.1 | NP_000031.1 | 4 | 443 | ||
GenBank mRNA | BC027977.1 | - | 4 | 441 | 5229117 | |
BC121081.1 | - | 4 | 443 | 40119733 | ||
BC134419.1 | - | 4 | 443 | 40119734 | ||
M28613.1 | - | 3 | 427 | |||
M28614.1 | - | 3 | 427 | |||
V01513.1 | - | 1 | 166 | |||
X00567.1 | - | 3 | 397 | |||
X01388.1 | - | 4 | 468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
