siRNA Details
Gene Symbol: KRT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000224.2 | NP_000215.1 | 2, 3 | 609 | ||
NM_199187.1 | NP_954657.1 | 3, 4 | 563 | |||
GenBank mRNA | AK129587.1 | - | 3, 4 | 485 | ||
AK223093.1 | - | 2, 3 | 551 | |||
AK308506.1 | - | 3, 4 | 548 | |||
AK313988.1 | - | 2, 3 | 552 | |||
AY762101.1 | - | 2, 3 | 498 | |||
BC000180.2 | - | 2, 3 | 535 | 2984862 | ||
BC004253.1 | - | 2, 3 | 520 | 3631540 | ||
BC008636.1 | - | 2, 3 | 533 | 3463780 | ||
BC020982.1 | - | 2, 3 | 543 | 3850724 | ||
BC072017.1 | - | 2, 3 | 539 | 6251574 | ||
BG753529.1 | - | 3, 4 | 563 | |||
BT019412.1 | - | 2, 3 | 494 | |||
CD106591.1 | - | 2, 3 | 609 | |||
X12881.1 | - | 2, 3 | 540 | |||
X12883.1 | - | 2, 3 | 476 |
Gene Symbol: KRT18P59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128036.1 | - | 1 | 903 |
Gene Symbol: PKNOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017018110.1 | XP_016873599.1 | 1 | 2067 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010664, CV561123, BC020474, M36376, AK167469, and others.
Gene Ontology Categories:
