siRNA Details
Gene Symbol: MTRF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114184.2 | NP_001107656.1 | 3 | 502 | ||
NM_001301047.2 | NP_001287976.1 | 3 | 502 | |||
NM_001301870.1 | NP_001288799.1 | 3 | 502 | |||
NM_001301871.1 | NP_001288800.1 | 2, 3 | 477 | |||
NM_019041.6 | NP_061914.3 | 3 | 502 | |||
NR_126056.1 | 2, 3 | 477 | ||||
GenBank mRNA | AK027684.1 | - | 2, 3 | 347 | ||
AK095240.1 | - | 3 | 412 | |||
AK225321.1 | - | 3 | 376 | |||
AK225970.1 | - | 3 | 376 | |||
AK290662.1 | - | 3 | 376 | |||
AK297671.1 | - | 3 | 477 | |||
BC011873.2 | - | 3 | 399 | 3029407 | ||
BC014428.1 | - | 3 | 388 | 4646472 | ||
BC105973.1 | - | 3 | 401 | 6049942 | ||
BX376520.2 | - | 2, 3 | 370 | |||
DA152954.1 | - | 3 | 502 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
