siRNA Details
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001016.3 | NP_001007.2 | 1 | 485 | ||
GenBank mRNA | BC002550.1 | - | 1570 | |||
BC017321.2 | - | 1 | 468 | 4565075 | ||
BC071930.1 | - | 1 | 464 | 6295016 | ||
BC095424.1 | - | 1 | 475 | 30792687 | ||
BQ276878.1 | - | 6 | 488 | |||
X53505.1 | - | 6 | 482 |
Gene Symbol: SNORA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096147.1 | - | 1 | 2975 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
