siRNA Details
Gene Symbol: MDFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300804.1 | NP_001287733.1 | 5, 6 | 826 | ||
NM_001300805.1 | NP_001287734.1 | 3, 4 | 506 | |||
NM_001300806.1 | NP_001287735.1 | 3, 4 | 775 | |||
NM_005586.3 | NP_005577.1 | 4, 5 | 689 | |||
XM_005249117.3 | XP_005249174.1 | 4, 5 | 692 | |||
XM_011514625.2 | XP_011512927.1 | 5, 6 | 977 | |||
XM_011514626.2 | XP_011512928.1 | 4, 5 | 800 | |||
XM_017010867.1 | XP_016866356.1 | 3, 4 | 1348 | |||
GenBank mRNA | AK290735.1 | - | 3, 4 | 773 | ||
BC007836.2 | - | 4, 5 | 638 | 4110841 | ||
BI824391.1 | - | 3, 4 | 486 | |||
BX397111.2 | - | 5, 6 | 809 | |||
U78313.1 | - | 4, 5 | 631 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
