siRNA Details
Gene Symbol: MDFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300804.1 | NP_001287733.1 | 5 | 726 | ||
NM_001300805.1 | NP_001287734.1 | 3 | 406 | |||
NM_001300806.1 | NP_001287735.1 | 3 | 675 | |||
NM_005586.3 | NP_005577.1 | 4 | 589 | |||
XM_005249117.3 | XP_005249174.1 | 4 | 592 | |||
XM_011514625.2 | XP_011512927.1 | 5 | 877 | |||
XM_011514626.2 | XP_011512928.1 | 4 | 700 | |||
XM_017010867.1 | XP_016866356.1 | 3 | 1248 | |||
GenBank mRNA | AK290735.1 | - | 3 | 673 | ||
BC007836.2 | - | 4 | 538 | 4110841 | ||
BG818571.1 | - | 4 | 588 | |||
BI824391.1 | - | 3 | 386 | |||
BX397111.2 | - | 5 | 709 | |||
U78313.1 | - | 4 | 531 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010783, NM_001276391, NM_001276390, NM_001109973, CX564914, and others.
Gene Ontology Categories:
