siRNA Details
Gene Symbol: LAMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105206.2 | NP_001098676.2 | 3 | 655 | ||
NM_001105207.2 | NP_001098677.2 | 3 | 672 | |||
NM_002290.4 | NP_002281.3 | 3 | 655 | |||
XM_005266983.3 | XP_005267040.2 | 3 | 528 | |||
XM_005266984.3 | XP_005267041.2 | 3 | 525 | |||
XM_017010854.1 | XP_016866343.1 | 3 | 525 | |||
GenBank mRNA | AB210027.1 | - | 3 | 507 | ||
AK304401.1 | - | 3 | 478 | |||
BC026237.1 | - | 3 | 487 | |||
BC066552.1 | - | 3 | 668 | 6164247 | ||
CR407622.1 | - | 2 | 257 | |||
S78569.1 | - | 3 | 447 | |||
U77706.1 | - | 3 | 612 | |||
X91171.1 | - | 3 | 540 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
