siRNA Details
Gene Symbol: LAMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105206.2 | NP_001098676.2 | 2 | 513 | ||
NM_001105207.2 | NP_001098677.2 | 2 | 530 | |||
NM_001105208.2 | NP_001098678.1 | 2 | 513 | |||
NM_001105209.2 | NP_001098679.1 | 2 | 530 | |||
NM_002290.4 | NP_002281.3 | 2 | 513 | |||
XM_005266983.3 | XP_005267040.2 | 2 | 386 | |||
XM_005266984.3 | XP_005267041.2 | 2 | 383 | |||
XM_017010854.1 | XP_016866343.1 | 2 | 383 | |||
GenBank mRNA | AB210027.1 | - | 2 | 365 | ||
AJ710266.1 | - | 2 | 386 | |||
AK304401.1 | - | 2 | 336 | |||
BC004241.1 | - | 2 | 344 | |||
BC026237.1 | - | 2 | 345 | |||
BC066552.1 | - | 2 | 526 | 6164247 | ||
BM014298.1 | - | 2 | 362 | |||
BP234809.1 | - | 2 | 513 | |||
BT006690.1 | - | 1 | 115 | |||
CR407622.1 | - | 1 | 115 | |||
DB271101.1 | - | 2 | 424 | |||
S78569.1 | - | 2 | 305 | |||
U77706.1 | - | 2 | 470 | |||
X91171.1 | - | 2 | 398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
