siRNA Details
Gene Symbol: BTNL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040462.2 | NP_001035552.1 | 4 | 1007 | ||
NM_001159707.1 | NP_001153179.1 | 3 | 659 | |||
NM_001159708.1 | NP_001153180.1 | 4 | 1007 | |||
NM_001159709.1 | NP_001153181.1 | 4 | 710 | |||
NM_001159710.1 | NP_001153182.1 | 3 | 510 | |||
NM_024850.2 | NP_079126.1 | 4 | 1007 | |||
XM_011534650.2 | XP_011532952.1 | 4 | 812 | |||
XM_011534651.2 | XP_011532953.1 | 3 | 1826 | |||
XM_011534652.2 | XP_011532954.1 | 3 | 1802 | |||
XM_011534653.2 | XP_011532955.1 | 2 | 363 | |||
GenBank mRNA | AK025111.1 | - | 4 | 925 | ||
AK225213.1 | - | 4 | 925 | |||
AK296597.1 | - | 4 | 665 | |||
AK298687.1 | - | 3 | 510 | |||
AK308012.1 | - | 3 | 495 | |||
AK309639.1 | - | 3 | 501 | |||
AY358523.1 | - | 4 | 955 | |||
BC119696.1 | - | 3 | 462 | 40116820 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
