siRNA Details
Gene Symbol: BTNL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040462.2 | NP_001035552.1 | 8 | 2038 | ||
NM_001159707.1 | NP_001153179.1 | 7 | 1690 | |||
NM_001159708.1 | NP_001153180.1 | 7 | 2147 | |||
NM_001159709.1 | NP_001153181.1 | 8 | 1741 | |||
NM_001159710.1 | NP_001153182.1 | 7 | 1541 | |||
NM_024850.2 | NP_079126.1 | 8 | 2168 | |||
XM_011534649.1 | XP_011532951.1 | 7 | 1924 | |||
XM_011534650.2 | XP_011532952.1 | 8 | 1843 | |||
XM_011534651.2 | XP_011532953.1 | 7 | 2857 | |||
XM_011534652.2 | XP_011532954.1 | 7 | 2833 | |||
XM_011534653.2 | XP_011532955.1 | 6 | 1394 | |||
GenBank mRNA | AK025111.1 | - | 8 | 2086 | ||
AY358523.1 | - | 8 | 1986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
