siRNA Details
Gene Symbol: DLEU2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002771.1 | 1 | 543 | |||
GenBank mRNA | AF254117.1 | - | 1 | 543 |
Gene Symbol: DLEU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002612.1 | 5 | 515 | |||
GenBank mRNA | AF264787.1 | - | 7 | 795 | ||
AF380424.1 | - | 5 | 694 | |||
AY455757.1 | - | LIT3420 DLEU2 |
8 | 804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
