siRNA Details
Gene Symbol: DLEU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002612.1 | 2, 3 | 337 | |||
GenBank mRNA | AF264787.1 | - | 3, 4 | 500 | ||
AF380424.1 | - | 2, 3 | 516 |
Gene Symbol: DLEU2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002771.1 | 1 | 250 | |||
GenBank mRNA | AF254117.1 | - | 1 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
