siRNA Details
Gene Symbol: ACYP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107.4 | NP_001098.1 | 2 | 119 | ||
NM_001302616.1 | NP_001289545.1 | 2 | 101 | |||
NM_001302617.1 | NP_001289546.1 | 2 | 297 | |||
NR_126393.1 | 2 | 119 | ||||
GenBank mRNA | AK312101.1 | - | 2 | 119 | ||
BC035568.1 | - | 2 | 104 | 5392248 | ||
BG614847.1 | - | 2 | 86 | |||
BP316920.1 | - | 2 | 82 | |||
CD675334.1 | - | 2 | 306 | |||
DB071115.1 | - | 2 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
