siRNA Details
Gene Symbol: LYZL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183058.2 | NP_898881.2 | 5 | 699 | ||
XM_011519306.2 | XP_011517608.1 | 4 | 663 | |||
GenBank mRNA | AF139543.1 | - | 5 | 678 | ||
BC066294.1 | - | 5 | 699 | 4374284 |
Gene Symbol: LYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032517.4 | NP_115906.3 | 5 | 743 | ||
GenBank mRNA | AY358694.1 | - | 5 | 688 | ||
BC021730.2 | - | 5 | 709 | 4825273 | ||
BG209290.1 | - | 2 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
