siRNA Details
Gene Symbol: LYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032517.4 | NP_115906.3 | 4 | 511 | ||
XM_005252627.3 | XP_005252684.1 | 4 | 521 | |||
GenBank mRNA | AY358694.1 | - | 4 | 456 | ||
BC021730.2 | - | 4 | 477 | 4825273 | ||
BP369317.1 | - | 4 | 522 |
Gene Symbol: LYZL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183058.2 | NP_898881.2 | 4 | 467 | ||
GenBank mRNA | AF139543.1 | - | 4 | 446 | ||
BC066294.1 | - | 4 | 467 | 4374284 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026092, AK006630, BC048450, AY572447, AK006241.
Gene Ontology Categories:
