siRNA Details
Gene Symbol: RPL21P28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026911.1 | 1 | 76 |
Gene Symbol: RPL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000982.3 | NP_000973.2 | 1 | 74 | ||
GenBank mRNA | AK130968.1 | - | 1 | 73 | ||
AK311795.1 | - | 1 | 71 | |||
AY320407.1 | - | 1 | 74 | |||
BC001603.1 | - | 1 | 56 | 3543702 | ||
BC007505.2 | - | 1 | 37 | 2964441 | ||
BC019002.2 | - | 1 | 29 | |||
BC062981.1 | - | 1 | 54 | 6084268 | ||
BC070184.1 | - | 1 | 58 | 4281405 | ||
BC070185.1 | - | 1 | 71 | |||
BC070323.1 | - | 1 | 71 | 4248655 | ||
BC070330.1 | - | 1 | 71 | 3931616 | ||
BC071902.1 | - | 1 | 54 | 4901492 | ||
BC104478.1 | - | 1 | 61 | |||
BC104667.1 | - | 1 | 75 | 6724053 | ||
BX647669.1 | - | 1 | 73 | |||
CR457032.1 | - | 1 | 31 | |||
U14967.1 | - | 1 | 60 | |||
U25789.1 | - | 1 | 64 | |||
X89401.1 | - | 1 | 44 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
