siRNA Details
Gene Symbol: TMEM242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018452.5 | NP_060922.2 | 4 | 2248 | ||
GenBank mRNA | AF217510.1 | - | 4 | 1588 | ||
AK000891.1 | - | 4 | 1497 | |||
BC029130.1 | - | 4 | 1513 | 5182086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
