siRNA Details
Gene Symbol: VPS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016516.2 | NP_057600.2 | 2, 3 | 307 | ||
XM_006712029.3 | XP_006712092.1 | 2, 3 | 275 | |||
GenBank mRNA | AF102177.1 | - | 1, 2 | 164 | ||
AK292669.1 | - | 3, 4 | 315 | |||
AY444798.1 | - | 1, 2 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
