siRNA Details
Gene Symbol: S100A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005620.1 | NP_005611.1 | 2 | 242 | ||
GenBank mRNA | AK312184.1 | - | 2 | 177 | ||
BC001410.2 | - | 2 | 202 | 3140092 | ||
BC014354.1 | - | 2 | 184 | 3685742 | ||
BT009912.1 | - | 1 | 122 | |||
D38583.1 | - | 2 | 242 | |||
D49355.1 | - | 2 | 209 | |||
D50374.1 | - | 2 | 138 | |||
EU794659.1 | - | 2 | 199 | |||
X80201.1 | - | 2 | 199 |
Gene Symbol: EIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA159721.1 | - | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
