siRNA Details
Gene Symbol: S100A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005620.1 | NP_005611.1 | 3 | 404 | ||
GenBank mRNA | AK312184.1 | - | 3 | 339 | ||
BC001410.2 | - | 3 | 364 | 3140092 | ||
BC014354.1 | - | 3 | 346 | 3685742 | ||
BT009912.1 | - | 2 | 284 | |||
D38583.1 | - | 3 | 404 | |||
D49355.1 | - | 3 | 371 | |||
D50374.1 | - | 3 | 300 | |||
EU794659.1 | - | 3 | 361 | |||
X80201.1 | - | 3 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
