siRNA Details
Gene Symbol: SFTPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098668.3 | NP_001092138.1 | 6 | 2067 | ||
NM_001320813.1 | NP_001307742.1 | 6 | 2070 | |||
NM_001320814.1 | NP_001307743.1 | 5 | 2136 | |||
XM_005270128.3 | XP_005270185.1 | 6 | 2048 | |||
XM_005270132.3 | XP_005270189.1 | 5 | 2035 | |||
XM_011540124.1 | XP_011538426.1 | 6 | 2163 | |||
XM_011540125.1 | XP_011538427.1 | 6 | 2106 | |||
XM_017016608.1 | XP_016872097.1 | 5 | 1959 | |||
GenBank mRNA | HQ021421.1 | - | 6 | 2016 | ||
HQ021422.2 | - | 6 | 2016 | |||
HQ021423.2 | - | 6 | 2016 | |||
HQ021424.1 | - | 6 | 2016 | |||
HQ021425.2 | - | 6 | 2016 | |||
HQ021426.1 | - | 6 | 2013 | |||
HQ021427.1 | - | 6 | 2013 | |||
HQ021428.2 | - | 6 | 2013 | |||
HQ021429.2 | - | 6 | 2013 | |||
HQ021430.1 | - | 6 | 2013 | |||
HQ021431.2 | - | 6 | 2013 | |||
HQ021432.1 | - | 6 | 2016 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
